Order Entry
Austria
ContactUsLinkComponent
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Certificates
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Certificates

Specifications

  • MDL No.:
    MFCD00236357
  • Physical form:
    Liquid

Specifications

Related Information

Frequently Bought Together