


(R)-(-)-2-Aminohexane ≥99%, ee 96%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70095-40-8
:
Formula:C₆H₁₅N
MDL Number:MFCD00671627
:101.19 g/mol
Boiling Pt:116 °C
Melting Pt:< –20 °C
Density:0.755
Flash Pt:–3 °C (26 °F)
UN:2733
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together