Potassium hydroxide 1 M (1 N) in ethanol volumetric solution, Fluka™
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:78 °C (1.013 hPa)
:0.88 g/cm3 (20 °C)
:11 °C
:Ambient
:215-181-3
:2924
:3,II
:GHS02
:GHS05
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together