di-Potassium hydrogen phosphate 1 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
:174.18 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-834-5
Frequently Bought Together