Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
di-Potassium hydrogen phosphate
di-Potassium hydrogen phosphate 1 mol/l in aqueous solution
di-Potassium hydrogen phosphate 1 mol/l in aqueous solution
di-Potassium hydrogen phosphate 1 mol/l in aqueous solution
786-487
:
G-Biosciences
:
di-Potassium hydrogen phosphate 1 mol/l in aqueous solution
786-487
:
G-Biosciences
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
174.18 g/mol
:
Ambient
:
231-834-5
:
7758-11-4
:
K₂HPO₄
:
MFCD00011383