
Okadaic acid 97%
Catalog # ACRO328370001
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 mg
- Seq:1286
Specifications
Specification Test Results
- Appearance:White to off-white Solid
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:805.02 g/mol
Melting Pt:164…166 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,II
:GHS06
CAS Number:78111-17-8
Formula:C₄₄H₆₈O₁₃
MDL Number:MFCD00083455