Cut resistant gloves, Camapur® Cut 620
Supplier: KCL by Honeywell
Certificates
About this item
These seamless HPPE lined gloves feature a knit wrist and a protective PU coating on the palm.
- PU coating ensures good grip - even when handling wet, slippery or slightly oily parts
- Elasticated knit wrist for wearing comfort
- Moisture-repellent, high mechanical resistance
- Free from the allergenic substances thiurames and dithiocarbamates
Ideal for assembly works with medium cut risks, metal sheet processing and the processing of synthetic material profiles.
Thickness: 1,2 ± 0,1 mm
Colour: White/grey
EN 388 (4342), Cat. II
Certifications: EN 388 (4342), Cat. II
Specifications
- Catalog No:
- KCHL620.7
- KCHL620.9
- KCHL620.8
- KCHL620.10
- KCHL620.11
- KCHL620.6
- Model:
- Cut 620
- Cut 620
- Cut 620
- Cut 620
- Cut 620
- Cut 620
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Finish:
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Coating on palm and fingers
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)
- EN 388 (4342)