ContactUsLinkComponent
Cleanroom hoods, for use with goggles, 5R181
Cleanroom hoods, for use with goggles, 5R181
Supplier:  FRISTADS KANSAS
Certificates
undefined
Cleanroom hoods, for use with goggles, 5R181
Supplier:  FRISTADS KANSAS
Specifications
  • Catalog No:
  • KWIN100649500
  • KWIN100649730
  • Model:
  • 5R181
  • 5R181
  • Garment application:
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Gender:
  • Unisex
  • Unisex
  • Fabric:
  • XR50 fabric (97% polyester and 3% conductive fibre)
  • XR50 fabric (97% polyester and 3% conductive fibre)
  • Weight:
  • Weight: 115 g/m²
  • Weight: 115 g/m²