Order Entry
Austria
ContactUsLinkComponent
DrySyn® heating blocks, DrySyn® Vortex
DrySyn® heating blocks, DrySyn® Vortex
Catalog # 442-0871
Supplier:  ASYNT
undefined
DrySyn® heating blocks, DrySyn® Vortex
Catalog # 442-0871
Supplier:  ASYNT

Specifications

  • Description:
    DrySyn Vortex, kit with 1 vortex overhead stirrer, 3 stirrers, 3 guides and 3×500 ml flask inserts
  • Seq:
    55
  • Pk:
    1

Specifications

About this item

For overhead stirring of three reactions using only one overhead stirrer.

  • Can be used with all standard hotplate stirrers or as stand-alone
  • Excellent stirring for viscous solutions
  • Compatible with DrySyn Multi system inserts, accommodates 3×100 ml, 3×250 ml and 3×500 ml flasks
  • Compatible with standard laboratory round bottom flasks