Distillation columns, Vigreux, with removable glass jacket
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket and NS cone, with removable glass jacket, horizontal and lateral indentations.
Frequently Bought Together