Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Insert Boxes

Storage Insert Boxes

Supplier:   hünersdorff GmbH
Storage Insert Boxes
Supplier:  hünersdorff GmbH
Certificates
Storage Insert Boxes

12 Options Available Below

Product Details & Documents

Insert boxes made of tough polystyrene (PS) for order in drawers, tool cupboards, storage cabinets and small parts boxes.

  • Colours suitable for foods
  • Suitable for assortment boxes and small parts cases

Ordering information: Transparent lids are available separately.

Product Family Options

Product Information

  • Colour
    W×D×H
    Material
    Availability
    Price
  • Red
    54×54×23 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Red
    54×54×63 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,00 €
    1,00 €
    1 * 1 ST
       1,00 €

  • Yellow
    54×108×23 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,10 €
    1,10 €
    1 * 1 ST
       1,10 €

  • Yellow
    54×108×63 mm
    PS
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST - 83,80 €
    83,80 €
    1 * 48 ST
       83,80 €

  • Blue
    108×108×23 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,60 €
    1,60 €
    1 * 1 ST
       1,60 €

  • Blue
    108×108×45 mm
    PS
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 71,20 €
    71,20 €
    1 * 40 ST
       71,20 €

  • Blue
    108×108×63 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,10 €
    6,10 €
    1 * 1 ST
       6,10 €

  • Green
    108×162×23 mm
    PS
    Log in for availability
    Log in for availability
    1 * 35 ST
    1 * 35 ST - 68,30 €
    68,30 €
    1 * 35 ST
       68,30 €

  • Green
    108×162×45 mm
    PS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 78,70 €
    78,70 €
    1 * 25 ST
       78,70 €

  • Green
    108×162×63 mm
    PS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3,90 €
    3,90 €
    1 * 1 ST
       3,90 €

  • Grey
    108×216×45 mm
    PS
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 74,70 €
    74,70 €
    1 * 20 ST
       74,70 €

  • Grey
    108×324×45 mm
    PS
    Log in for availability
    Log in for availability
    1 * 15 ST
    1 * 15 ST - 65,00 €
    65,00 €
    1 * 15 ST
       65,00 €

Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 324×108 mm
    Lid for insert box, PS, 324×108 mm
    HUEN253000
    Clear
    Log in for availability
    1 * 10 ST

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 216×108 mm
    Lid for insert box, PS, 216×108 mm
    HUEN252900
    Clear
    Log in for availability
    1 * 10 ST - 20,60 €

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 162×108 mm
    Lid for insert box, PS, 162×108 mm
    HUEN252800
    Clear
    Log in for availability
    1 * 1 ST - 2,10 €

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 108×108 mm
    Lid for insert box, PS, 108×108 mm
    HUEN252700
    Clear
    Log in for availability
    1 * 1 ST - 1,90 €

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 108×54 mm
    Lid for insert box, PS, 108×54 mm
    HUEN252600
    Clear
    Log in for availability
    1 * 1 ST - 1,50 €

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Lid for insert box, PS, 54×54 mm
    Lid for insert box, PS, 54×54 mm
    HUEN252500
    Clear
    Log in for availability
    1 * 1 ST - 1,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.