Orders LinkContactUsLinkComponent
Teaching camera
Teaching camera
Digital teaching camera, 3 MP
Catalog # DIDAPOF010810
Supplier:   DIDALAB
Teaching camera
Catalog # DIDAPOF010810
Supplier:  DIDALAB
Teaching camera
Teaching camera
Digital teaching camera, 3 MP
Price
1 134,87 €
Log in for availability
1 * 1 ST
Product Details & Documents
The pedagogical model of the digital camera can be put on a Ø-10 mm rod or on a camera’s stand.
  • Pentax lenses: One with a fixed 50-mm focus, the other with a 28 to 70-mm focus
  • Focus, aperture, time exposure, field depth, magnification, sensor, sensibility and accuracy
  • Image reflection with mirror to understand the Reflex aiming
By setting the lens, the students can understand the field depth and the digital aperture principle.
Delivery information: Supplied with specific software and a complete manual.
Specifications
  • Resolution:
    3 MP
  • Description:
    Digital teaching camera