Capillary columns, TracePLOT™ TG-BOND Q
Supplier: Thermo Fisher Scientific
Certificates
About this item
Non-polar columns for oxygenated compounds and solvents.
- Non-polar 100% divinyl benzene phase
- Particles incorporated into the walls of the tubing for essentially no particle release
Frequently Bought Together