ContactUsLinkComponent
CCD camera, POF 010 300
CCD camera, POF 010 300
Catalog # DIDAPOF010300
Supplier:  DIDALAB
undefined
CCD camera, POF 010 300
Catalog # DIDAPOF010300
Supplier:  DIDALAB
Specifications
  • Description:
    Basic CCD camera
  • Pk:
    1
Specifications
About this item
This Caliens camera enables measuring very precise distances.
  • CCD sensor, 2048 pixels
  • User-friendly software, Windows compatible
  • Power supplied via USB port (connected to a PC)
Delivery information: Supplied with Ø 10 mm rod for use with optical benches, software and USB cable.