ContactUsLinkComponent
Visual comparator test kit for dissolved oxygen by the indigo carmine method, CHEMets
Visual comparator test kit for dissolved oxygen by the indigo carmine method, CHEMets
Catalog # CHETK-7512
Supplier:  CHEMetrics
CAS Number:  
undefined
Visual comparator test kit for dissolved oxygen by the indigo carmine method, CHEMets
Catalog # CHETK-7512
Supplier:  CHEMetrics
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Dissolved oxygen kit, indigo carmne method
  • Range:
    1-2-3-4-5-6-8-10-12 ppm
Specifications
About this item
Test kits for environmental and drinking water applications in the ppm range.
  • Visual indication of dissolved oxygen by formation of blue product
  • Method is not to interferences from temperature, salinity, or dissolved gases such as sulfide
  • Results are expressed as ppm (mg/L) O₂
Delivery: Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, comparator, 25 ml sample cup and instructions