pH buffer solutions
Supplier: Hach
Certificates
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Water Sampling Bottles, Sterile
Supplier: VWR Collection
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Sodium sulphite, anhydrous 95.0-100.5% Ph. Eur.
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetic acid glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Conductivity standards
Supplier: Hach
Inoculating loops, PS, calibrated
Supplier: DELTALAB