Sealing film, self-adhesive, for cold storage
Catalog # 731-0441
Supplier: Brand
Specifications
- Description:Microplate sealing film, self-adhesive, for cold storage, aluminium, single sheets
- Pk:100
Specifications
About this item
Aluminium sealing film can be pierced easily with a pipette tip for sample removal. Temperature range: −80 to +120 °C.