ContactUsLinkComponent
Insulating Boxes for TP 5 P Including Bottles/Containers
Insulating Boxes for TP 5 P Including Bottles/Containers
Catalog # 701-0084
Supplier:  MAXX
undefined
Insulating Boxes for TP 5 P Including Bottles/Containers
Catalog # 701-0084
Supplier:  MAXX
Specifications
  • Description:
    Insulating box, 24×1000 ml
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
The insulating boxes in the table below each have an integral distributor for the relevant bottle configuration. The TP 5 P sampler is simply mounted on the lid of the insulating box.
Ordering information: For details of insulating boxes for other container sizes and for active cooling options please visit the website or contact your local Avantor sales office.