ContactUsLinkComponent
VWR® Low Flow Cells for pH Measurements
VWR® Low Flow Cells for pH Measurements
  662-0457
undefined
VWR® Low Flow Cells for pH Measurements
  662-0457
  • Description:
    Low flow cell 200 µl, acrylic
  • Seq:
    32
  • Pk:
    1
VWR® low flow cells are designed for use in a variety of instrumental techniques such as FIA, HPLC and IEF. They are also used to enable a continuous monitoring of the pH at low volume flows in bench scale chemical systems. They are supplied with inlet and outlet fittings sized for standard ¹/₁₆' or 1,6 mm OD tubing.
  • Designed for clean, sweeping flow at low volume
  • Designed for use with VWR® large surface flat tip electrode Cat. No. 662-0456