ContactUsLinkComponent
Standards for gas chromatography, OQ/PV headspace sample standard
Standards for gas chromatography, OQ/PV headspace sample standard
Catalog # HEWL5182-9733
Supplier:  Agilent
CAS Number:  
Standards for gas chromatography, OQ/PV headspace sample standard
Catalog # HEWL5182-9733
Supplier:  Agilent
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    OQ/PV headspace sample standard
Specifications
About this item
OQ/PV headspace sample standard contains 0,2 to 0,3% t-butyl disulphide, 1,2-dichlorobenzene, and nitrobenzene in ethanol.