Orders LinkContactUsLinkComponent
VWR® Mr. Lab, Chairs and Stools
VWR® Mr. Lab, Chairs and Stools
   Sustainable Options Available
Supplier:   VWR Collection
VWR® Mr. Lab, Chairs and Stools
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR® Mr. Lab, Chairs and Stools
11 Options Available Below
Product Details & Documents
All chairs and stools are available with a five star base which is made from aluminium.
  • Chair backrest adjustable in height and inclination
  • Seat height adjustable by gas-lift
  • Aluminium five star base
The perfect combination of comfort and ergonomics for optimal performance. Anatomically-moulded cushions with pelvis and integrated lumbar support. Very comfortable, with large, soft upholstery in artificial leather covering, washable and disinfectant-proof.
Glides: The glides are wear-proof plastic and have a large surface area offering a high level of stability.
Castors: Dual-wheel castors with load-sensitive brakes have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors these chairs are only mobile when no one is sitting on them.
Warranty: 10-year long-term guarantee, free shipping of spare parts during the whole period of validity.
Certifications: Conform to security level S2, GMP.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 132-0416
  • 132-0939
  • 132-0417
  • 132-0501
  • 132-0965
  • 132-0966
  • 132-0967
  • 132-0425
  • 132-0426
  • 132-0963
  • 132-0964
  • Environmentally Preferable:
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • Environmental certification:
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
  • ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
Specifications
Product Family Options
Chairs
Product Information
  • Height
    Foot style
    Foot ring
    Availability
    Price
  • 132-0416
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    470 - 620 mm
    Castors
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
    1 * 1 ST
       368,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0416
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0939
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    470 - 620 mm
    Castors, "Stop&Go"
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 381,00 €
    381,00 €
    1 * 1 ST
       381,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0939
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0417
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    600 - 870 mm
    Glides
    +
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 472,00 €
    472,00 €
    1 * 1 ST
       472,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0417
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0501
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from blue skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    640 - 910 mm
    Castors, "Stop&Go"
    +
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 488,00 €
    488,00 €
    1 * 1 ST
       488,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0501
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0965
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    470 - 620 mm
    Soft castors for hard floors
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 368,00 €
    368,00 €
    1 * 1 ST
       368,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0965
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0966
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    600 - 870 mm
    Large glides
    Height adjustable, chrome plated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 472,00 €
    472,00 €
    1 * 1 ST
       472,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0966
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0967
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    Laboratory chair, made from black skai, with permanent contact backrest and aluminium polished base, plastic parts in black
    620 - 890 mm
    Castors, "Stop&Go"
    Height adjustable, chrome plated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 488,00 €
    488,00 €
    1 * 1 ST
       488,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0967
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
Stools
Product Information
  • Height
    Foot style
    Foot ring
    Availability
    Price
  • 132-0425
    Laboratory stool, plastic black frame, seat made from black skai; height adjustment based on pneumatic spring (plastic and steel part are black)
    Laboratory stool, plastic black frame, seat made from black skai; height adjustment based on pneumatic spring (plastic and steel part are black)
    460 - 630 mm
    Castors
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 175,00 €
    175,00 €
    1 * 1 ST
       175,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0425
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0426
    Laboratory stool, plastic black frame, seat made from black PU foam; height adjustment based on pneumatic spring (plastic and steel part are black)
    Laboratory stool, plastic black frame, seat made from black PU foam; height adjustment based on pneumatic spring (plastic and steel part are black)
    460 - 630 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 178,00 €
    178,00 €
    1 * 1 ST
       178,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0426
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0963
    Laboratory stool, extra large seat (Ø 400 mm) made from black skai leather, PA base (plastic and steel part are black)
    Laboratory stool, extra large seat (Ø 400 mm) made from black skai leather, PA base (plastic and steel part are black)
    600 - 870 mm
    Castors, "Stop&Go"
    Height adjustable, chrome plated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 254,00 €
    254,00 €
    1 * 1 ST
       254,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0963
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
  • 132-0964
    Laboratory stool, extra large seat (Ø 350 mm) made from PU foam, PA base (plastic and steel part are black)
    Laboratory stool, extra large seat (Ø 350 mm) made from PU foam, PA base (plastic and steel part are black)
    600 - 870 mm
    Castors, "Stop&Go"
    Height adjustable, chrome plated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
    Specifications:
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    132-0964
    Environmentally Preferable
    Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
    Environmental certification
    ISO 14001 (No. 20240410), EMAS (No. DE-168-00006), ISO 50001 (No.Z030-2023-01), FSC Certified Wood (No. SGSCH-COC-009712), Öko-tex-100 (No. Sh015 25042 or No. 11-43686)
      Sustainable Materials|Low Manufacturing Impact|Safer Human and Environmental Health
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Castors, hard for soft floors, 1 set of 5 castors
    Castors, hard for soft floors, 1 set of 5 castors
    132-0427
    Log in for availability
    1 * 1 SET - 40,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Armrests, black
    Armrests, black
    132-0423
    Log in for availability
    1 * 2 ST - 84,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Height adjustable foot ring (compatible with all models, including All-In-One and Mr. Lab chairs)
    Height adjustable foot ring (compatible with all models, including All-In-One and Mr. Lab chairs)
    139-0215
    Log in for availability
    1 * 1 ST - 91,20 €