ContactUsLinkComponent
Microorganism tests, BART test for acid producing bacteria
Microorganism tests, BART test for acid producing bacteria
Artikelnr. HACH2831409
Supplier:  Hach
CAS Number:  
undefined
Microorganism tests, BART test for acid producing bacteria
Artikelnr. HACH2831409
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    9
  • Description:
    BART test for acid producing bacteria
Specifications
About this item
BART biodetectors are excellent diagnostic tools to help identify the presence and activity of various bacteria.
  • Simple yet effective method for monitoring the population size and/or activity of specific groups of bacteria
  • Easy to use, requiring no elaborate or costly equipment and no specialized training
  • Effective and affordable tests are easy to interpret and can be performed at room temperature in virtually any environment