Competent cells, expression strains, HMS174
About this item
A common method for transformation of plasmids into E.coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Every Novagen® competent cell strain is verified for phenotype and purity, and is guaranteed for transformation efficiency.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilising pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0,2 ml, which provides enough cells for ten transformations. All kits are supplied with SOC medium and test plasmid. Several strains are also available as Singles™ Competent Cells, which are provided in single-use, 50 μl volumes for extra convenience and efficiency.
HMS174 strains provide the recA mutation in a K-12 background. Like BLR, these strains may stabilise certain target genes whose products may cause the loss of the DE3 prophage.
5 Options Available Below
Product Details & Documents
A common method for transformation of plasmids into E.coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Every Novagen® competent cell strain is verified for phenotype and purity, and is guaranteed for transformation efficiency.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilising pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0,2 ml, which provides enough cells for ten transformations. All kits are supplied with SOC medium and test plasmid. Several strains are also available as Singles™ Competent Cells, which are provided in single-use, 50 μl volumes for extra convenience and efficiency.
HMS174 strains provide the recA mutation in a K-12 background. Like BLR, these strains may stabilise certain target genes whose products may cause the loss of the DE3 prophage.
Product Family Options
Product Information
- AvailabilityPrice
- 69452-3HMS174 competent cells, 2×200 μl
- 69452-4HMS174 competent cells, 5×200 μl
- 69453-3HMS174(DE3) competent cells, 2×200 μl
- 69453-4HMS174(DE3) competent cells, 5×200 μl
- 70234-3HMS174 competent cell set, HMS174, HMS174(DE3), HMS174(DE3)pLysSHMS174 competent cell set, HMS174, HMS174(DE3), HMS174(DE3)pLysSLog in for availability1 * 1,2 mL425,00 €
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



