ContactUsLinkComponent
Flame-retardant bomber jacket with detachable sleeves, high visibility, Oregon 1874
Flame-retardant bomber jacket with detachable sleeves, high visibility, Oregon 1874
Supplier:  SIOEN
Certificates
undefined
Flame-retardant bomber jacket with detachable sleeves, high visibility, Oregon 1874
Supplier:  SIOEN
Specifications
  • Catalog No:
  • SIOE1874/278/S
  • SIOE1874/278/M
  • SIOE1874/278/L
  • SIOE1874/278/3XL
  • SIOE1874/278/1XL
  • SIOE1874/278/2XL
  • Model:
  • Oregon 1874
  • Oregon 1874
  • Oregon 1874
  • Oregon 1874
  • Oregon 1874
  • Oregon 1874
  • Garment application:
  • Fire protection, Waste water
  • Fire protection, Waste water
  • Fire protection, Waste water
  • Fire protection, Waste water
  • Fire protection, Waste water
  • Fire protection, Waste water
  • Risk area:
  • Flame-retardant, Anti-static
  • Flame-retardant, Anti-static
  • Flame-retardant, Anti-static
  • Flame-retardant, Anti-static
  • Flame-retardant, Anti-static
  • Flame-retardant, Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • Polyester/PU
  • Polyester/PU
  • Polyester/PU
  • Polyester/PU
  • Polyester/PU
  • Polyester/PU
  • Weight:
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²