Orders LinkContactUsLinkComponent
Reference electrode filling solutions
Reference electrode filling solutions
Reference electrode filling solutions
Supplier:  Mettler - Toledo
Certificates
Reference electrode filling solutions
Reference electrode filling solutions
9 Options Available Below
Product Details & Documents
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 662-2886
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.)
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.)
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 12,90 €
    12,90 €
    1 * 25 mL
       12,90 €
  • METT51350076
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51350084
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml
    FRISCOLYT-B®, for measurement at low temperature and for media with organic compounds (oil, proteins etc.), 250 ml
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 105,00 €
    105,00 €
    1 * 6 ST
       105,00 €
  • METT51343180
    Potassium chloride (KCl) 3 mol/l, for ARGENTHAL™ reference systems
    Potassium chloride (KCl) 3 mol/l, for ARGENTHAL™ reference systems
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 9,70 €
    9,70 €
    1 * 25 mL
       9,70 €
  • METT51350072
    Potassium chloride (KCl) 3 mol/l, for ARGENTHAL™ reference systems
    Potassium chloride (KCl) 3 mol/l, for ARGENTHAL™ reference systems
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51343184
    Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/AgCl reference systems
    Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/AgCl reference systems
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 12,90 €
    12,90 €
    1 * 25 mL
       12,90 €
  • METT51350082
    Potassium chloride (KCl) 3 mol/l, AgCI saturated, for Ag/AgCl reference systems, 250 ml bottle
    Potassium chloride (KCl) 3 mol/l, AgCI saturated, for Ag/AgCl reference systems, 250 ml bottle
    6
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 103,00 €
    103,00 €
    1 * 6 ST
       103,00 €
  • METT51350074
    Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/Cl reference systems
    Potassium chloride (KCl) 3 mol/l, AgCl saturated, for Ag/Cl reference systems
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 27,80 €
    27,80 €
    1 * 250 mL
       27,80 €
  • METT51343182
    Potassium nitrate (KNO₃) 1 mol/l, bridge electrolyte, for DX series ISE half-cells
    Potassium nitrate (KNO₃) 1 mol/l, bridge electrolyte, for DX series ISE half-cells
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 12,90 €
    12,90 €
    1 * 25 mL
       12,90 €