Order Entry
Austria
Orders LinkContactUsLinkComponent
pH Electrode storage solution

pH Electrode storage solution

Potassium chloride (KCl) 3 M, 500 mL
Catalog # 705-1030
Supplier:   Hach
pH Electrode storage solution
Catalog # 705-1030
Supplier:  Hach
CAS Number:  
pH Electrode storage solution
pH Electrode storage solution

Potassium chloride (KCl) 3 M, 500 mL

CAS Number:  
Price
55,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Potassium chloride (KCl) 3 M

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.