pH buffer solutions, colour coded
Supplier: Hach
Jump to:
About this item
For calibration of pH meters
- Colour coded for easy identification
- Accuracy (pH): ±0,02 at 25 °C
Certifications: Traceable to NIST.
12 Options Available Below
Product Details & Documents
For calibration of pH meters
- Colour coded for easy identification
- Accuracy (pH): ±0,02 at 25 °C
Certifications: Traceable to NIST.
Product Family Options
Individual pH buffer solutions
Product Information
- pH valuepH accuracypH buffer typeColourReference temperaturePkAvailabilityPrice
- HACH2283426Buffer solution
- HACH2283436Buffer solution
- HACH2283449.99Buffer solution
- HACH2283461Buffer solution
- HACH2283526Buffer solution
- HACH2283536Buffer solution
- HACH2283549.99Buffer solution
- HACH2283561Buffer solution
- HACH2283626Buffer solution
- HACH2283649.99Buffer solution
- HACH2283661Buffer solution
Buffer kit
Product Information
- pH valuepH accuracypH buffer typeColourReference temperaturePkAvailabilityPrice
- 705-1029Buffer solution kit, pH 4,01 (red)/7,00 (yellow)/10,01 (blue), 500 ml eachBuffer solution kit, pH 4,01 (red)/7,00 (yellow)/10,01 (blue), 500 ml each4,01/7,00/10,01-1 KITLog in for availability1 * 1 KIT52,30 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



