ContactUsLinkComponent
Laboratory glass cleaner, borer 24 FORTE
Laboratory glass cleaner, borer 24 FORTE
Catalog # 148-5219
Supplier:  Borer Chemie
undefined
Laboratory glass cleaner, borer 24 FORTE
Catalog # 148-5219
Supplier:  Borer Chemie
Specifications
  • Description:
    borer 24 FORTE
  • Pk:
    10 kg
Specifications
About this item
Highly alkaline cleaning powder for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinseable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum
  • Perfect results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Contains active chlorine as a cleaning booster
  • EDTA-, NTA- and surfactant-free, no release of surfactants into waste water
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process