ContactUsLinkComponent
A-1 medium broth MPN tubes
A-1 medium broth MPN tubes
  HACH2560915
 :  Hach
 :  
undefined
A-1 medium broth MPN tubes
  HACH2560915
 :  Hach
 :  
  • Pk:
    15
  • Standards:
    -
  • Description:
    A-1 medium broth MPN tubes
A-1 Medium broth tubes provide detection of faecal coliform bacteria using the Most Probable Number (MPN) method.
  • Save time and labor costs
  • Minimal training requirements