Cut resistant gloves, ActivArmr® 70-765 / 70-766
Supplier: Ansell
Certificates
About this item
These gloves knitted from nylon, acrylic and Kevlar® fibres feature a premium leather coating on the palm.
- Reinforced thumb and seams relocated to the backs of the fingers: Reduce exposure to cuts and offer greater protection and longer life
- Gloves guard against minor injuries and punctures
- Superior grip even in oily environments thanks to the premium leather coating
- 70-766 version has a longer cuff for additional protection
- Plaited inside for comfort with coloured polyester/cotton
Ideal for press and stamping applications, handling heavy and oily metal pieces and for handling sharp or irregular-edged metal parts.
Certifications: EN 388:2016 (4X43D), EN 407 (X1XXXX); Cat. III
Specifications
- Catalog No:
- ANSE70-765/9
- ANSE70-765/8
- ANSE70-765/11
- ANSE70-765/10
- ANSE70-766/7
- ANSE70-766/9
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Nylon/Kevlar®
- Finish:
- Leather palm
- Leather palm
- Leather palm
- Leather palm
- Leather palm
- Leather palm
- Gauge:
- 10
- 10
- 10
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)