Sample tubes for stool, with screw cap
About this item
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
Clair tube with overlapping screw cap for optimal hygiene when opening, reliably sealed with long-lasting spoon for about bean-sized samples.
Certifications: Conforms to DIN EN 829 for the dispatch of stool samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
6 Options Available Below
Product Details & Documents
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
Clair tube with overlapping screw cap for optimal hygiene when opening, reliably sealed with long-lasting spoon for about bean-sized samples.
Certifications: Conforms to DIN EN 829 for the dispatch of stool samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
- CapacityVersionCap colourExt.ØHeightPackedAvailabilityPrice
- 13 mlmounted, sterileBrown16 mm100 mm100/1000 (bag/ctn)Log in for availability1 * 1.000 ST328,00 €