Orders LinkContactUsLinkComponent
Plugs for test tubes
Plugs for test tubes
Supplier:   BOETTGER PAUL
Plugs for test tubes
Supplier:  BOETTGER PAUL
Certificates
Plugs for test tubes
Plugs for test tubes
9 Options Available Below
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
  • Not autoclavable
  • Easy handling with a high, ribbed grip
  • Easy to open by tilting 'one-hand operation'
Product Family Options
Product Information
  • Material
    Size
    Colour
    Availability
    Price
  • 12 mm
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 17,10 €
    17,10 €
    1 * 1.000 ST
       17,10 €
  • 12 mm
    Blue
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 18,20 €
    18,20 €
    1 * 1.000 ST
       18,20 €
  • 12 mm
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 18,20 €
    18,20 €
    1 * 1.000 ST
       18,20 €
  • 12 mm
    Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 18,20 €
    18,20 €
    1 * 1.000 ST
       18,20 €
  • 12 mm
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 18,20 €
    18,20 €
    1 * 1.000 ST
       18,20 €
  • LDPE
    16 mm
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 21,40 €
    21,40 €
    1 * 1.000 ST
       21,40 €
  • 16 mm
    Red
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 23,20 €
    23,20 €
    1 * 1.000 ST
       23,20 €
  • LDPE
    16 mm
    Yellow
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 23,20 €
    23,20 €
    1 * 1.000 ST
       23,20 €
  • 16 mm
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 23,20 €
    23,20 €
    1 * 1.000 ST
       23,20 €