Order Entry
Austria
ContactUsLinkComponent
Affinity coupling media, EAH Sepharose™ 4B
Affinity coupling media, EAH Sepharose™ 4B
Catalog # 17-0569-01
Supplier:  Cytiva
CAS Number:  
undefined
Affinity coupling media, EAH Sepharose™ 4B
Catalog # 17-0569-01
Supplier:  Cytiva
CAS Number:  

Specifications

  • Particle Size:
    90 µm
  • Pk:
    50 ml
  • Description:
    EAH Sepharose™ 4B
  • Matrix:
    Agarose, 4%
  • Storage temperature:
    4 - 8 °C
  • Flow rate:
    <75 cm/h
  • Type:
    Liquid

Specifications

About this item

EAH Sepharose™ 4B allows for the stable carbodiimide-based coupling of carboxyl groups to Sepharose® 4B via an 11-atom hydrophilic spacer arm that permits extremely stable coupling.

Stable carbodiimide-based coupling of carboxyl groups to 4B via an 11-atom hydrophilic spacer arm permits very stable coupling. Stable to all commonly used aqueous Sepharose buffers.