Order Entry
Austria
ContactUsLinkComponent
 
VWR® ARISTAR®, Multi-Element Quality Control Standard 1 mg/L, 28 Elements in Nitric Acid 5% for ICP
Catalog # 05200.185
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard 1 mg/L, 28 Elements in Nitric Acid 5% for ICP
Catalog # 05200.185
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 1 mg/l {Al; Ag; As; B; Ba; Be; Bi; Ca; Cd; Co; Cr; Cu; Fe; K; Li; Mg; Mn; Mo; Na; Ni; Pb; Sb; Se; Sr; Ti; Tl; V; Zn} in nitric acid 5% ARISTAR® for ICP

Specifications

About this item

28 elements 1 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.