Stainless steel semi-prep frits, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Qualitative filter paper, standard grades MN 615 and 615 1/4, circles, sheets and folded filters
Supplier: MACHEREY-NAGEL
Titration beakers
Supplier: Mettler - Toledo
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Flasks, round bottom, with two necks and standard ground joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW