Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 32 Elements in 5% Nitric Acid, for ICP
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 32 Elements in 5% Nitric Acid, for ICP
Catalog # 89186.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard, 100 mg/L for 32 Elements in 5% Nitric Acid, for ICP
Catalog # 89186.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 100 mg/L {Ag; Al; Ba; Bi; Ca; Cd; Co; Cr; Cu; Fe; Ga; Ge; In; K; Li; Mg; Mn; Mo; Na; Nb; Ni; P; Pb; Re; Sb; Si; Sn; Ta; Ti; V; W; Zn} in nitric acid 5% ARISTAR® for ICP

Specifications

About this item

32 Elements 100 mg/L: Ag, Al, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, Ge, In, K, Li, Mg, Mn, Mo, Na, Nb, Ni, P, Pb, Re, Sb, Si, Sn, Ta, Ti, V, W, Zn

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.