Order Entry
Austria
ContactUsLinkComponent
VWR® ARISTAR®, ICP Multi-Element Quality Control Standard 10 mg/L, 22 Elements in Nitric Acid 5%
VWR® ARISTAR®, ICP Multi-Element Quality Control Standard 10 mg/L, 22 Elements in Nitric Acid 5%
Catalog # 88724.150
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, ICP Multi-Element Quality Control Standard 10 mg/L, 22 Elements in Nitric Acid 5%
Catalog # 88724.150
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Description:
    Multi-element quality control standard 10 mg/L {As; Ba; Be; Cd; Co; Cr; Cu; Fe; Al; Mn; Mo; Ni; Pb; Sb; Se; Sn; Ti; Tl; V; U; Te; Zn} in nitric acid 5% ARISTAR® for ICP

Specifications

About this item

22 elements 10 mg/L: As, Ba, Be, Cd, Co, Cr, Cu, Fe, Al, Mn, Mo, Ni, Pb, Sb, Se, Sn, Ti, Tl, V, U, Te, Zn.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.