ContactUsLinkComponent
Holder for round bottom flasks
Holder for round bottom flasks
Catalog # AZLOTWR038
Supplier:  SciLabware
undefined
Holder for round bottom flasks
Catalog # AZLOTWR038
Supplier:  SciLabware
Specifications
  • Ø top:
    145 mm
  • Pk:
    1
Specifications
About this item
This stand is suitable for most round bottomed flasks.
  • Good chemical resistance
  • Stepped concentric rings to help stabilise flask