ContactUsLinkComponent
Sedimentation cones, Imhoff type, Azlon®
Sedimentation cones, Imhoff type, Azlon®
Catalog # AZLOSWX008
Supplier:  SciLabware
undefined
Sedimentation cones, Imhoff type, Azlon®
Catalog # AZLOSWX008
Supplier:  SciLabware
Specifications
  • Type:
    bottom end with stopper
  • Description:
    Imhoff cones 1000 ml
  • Pk:
    1
Specifications
About this item
Colourless cone, made up of transparent styrene-acrylonitrile S.A.N polymer.
  • With moulded graduations
  • Stopper at the bottom facilitates the removal of sample and cleaning
Certifications: Complies to DIN 12672