ContactUsLinkComponent
Closed top caps for storage vials
Closed top caps for storage vials
Catalog # NSCAB7815-20
undefined
Closed top caps for storage vials
Catalog # NSCAB7815-20
Specifications
  • Septum:
    PTFE
  • Description:
    Closed top cap, 20-400, PP
  • Seq:
    05
  • Pk:
    100
Specifications
About this item
PTFE on film/foam backing offers broad chemical resistance.
Not autoclavable.