Volumetric flasks, class A, KIMAX®, KIMBLE®
KIML28014-100
- Capacity:100 ml
- Height:180 mm
- Seq:05
- Pk:12
- Tolerance:± 0,08 ml
Manufactured from 33 expansion, low extractable borosilicate glass with standard taper glass stopper.
- Calibrated to contain
- With pennyhead glass stopper
- Flasks feature a single graduation ring blasted on the neck and a marking spot
: Sizes 5 ml and larger are designed from ASTM Specification E288, Class A requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.