ContactUsLinkComponent
Luer-to-MicroTight® adapter, Upchurch Scientific®
Luer-to-MicroTight® adapter, Upchurch Scientific®
Catalog # 554-3142
Luer-to-MicroTight® adapter, Upchurch Scientific®
Catalog # 554-3142
Specifications
  • Description:
    P-662 Luer-to-MicroTight® adapter assembly, 0,150 mm thru-hole
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
PEEK. Ideal for infusing samples into lab-on-a-chip devices. This product introduces only 14 nl of additional volume to the flow path. Use it to directly connect a Luer-tip syringe or other product that terminates with a standard male Luer to 360 µm ext. Ø capillary tubing without tubing sleeves. MicroTight® fittings are included.
  • Pressure rating up to 45 psi (2,4 bar)