Accessories for Scan® 100
Supplier: INTERSCIENCE
Certificates
About this item
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Universal Filter Tips
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sulphanilamide ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
DURAN® Test Tubes with or without Rim
Supplier: DWK Life Sciences
pH electrodes, SenTix®
Supplier: WTW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Sterilised filter bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
UV bench lamps, XX-series
Supplier: UVP ULTRA VIOLET PRODUCTS
VWR®, Environmental 90 mm Petri plates
Supplier: VWR Chemicals