Dual/Triple scale handheld refractometers, MASTER series
: Atago
The MASTER refractometers are designed for clinical applications.
- Plastic design resistant to salty and acid samples
- The (alpha) models include ATC and are IP 65 rated except eyepiece
- Ergonomic grip and prism housing
Samples cover the prism without having to lift the daylight plate.
- Catalog No:
- ATAG2771
- ATAG2773
- ATAG2793
- ATAG2791
- Model:
- MASTER-URC/Nα
- MASTER-URC/NM
- MASTER-URC/NM
- MASTER-URC/Nα
- Sample size:
- 1 drop
- 1 drop
- 1 drop
- 1 drop
- Accuracy:
- ±0,0005 nD
- ±0,0005 nD
- ±0,001 nD
- ±0,001 nD
- Range:
- Urine SG 1,000 - 1,050; Serum protein 0,0 - 12,0 g/100 ml ; 1,3330 - 1,3660 nD
- Urine SG 1,000 - 1,050; Serum protein 0,0 - 12,0 g/100 ml ; 1,3330 - 1,3660 nD
- Urine SG 1,000 - 1,050
- Urine SG 1,000 - 1,050 ; 1,333 - 1,356 nD
- Resolution:
- 0,001 SG/0,0005 nD/ 0,2 g/100 ml
- 0,001 SG/0,0005 nD/ 0,2 g/100 ml
- 0,001 SG/nD
- 0,001 SG/nD
- Weight:
- 110 g
- 110 g
- 110 g
- 110 g
Frequently Bought Together