0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Mops
Accessories for Ultraspeed Mop System
Accessories for Ultraspeed Mop System
129-0928
:
FHP VILEDA SCS
Accessories for Ultraspeed Mop System
129-0928
:
FHP VILEDA SCS
Description:
Colour clip, red (10×1×1 cm)
Seq:
01
Pk:
1