Ultrasonic baths, BactoSonic®
Bactosonic gently removes the biofilms from implant surfaces.
- Works with very low ultrasound intensity
- Biofilm is removed without killing bacteria
- Compare to standard methods up to 10000 times more bacteria can be detected
: Complete set includes ultrasonic bath, wire frame for foil test, TICKOPUR TR-3 concentrate (250 ml), implant boxes, and box trays.
- Catalog No:
- BAND3291
- BAND3290
- Model:
- BactoSonic® 14.2
- BactoSonic® 14.2
- Capacity:
- 9,5 L
- 9,5 L
- Frequency:
- 40 kHz
- 40 kHz
- Tank W×D×H:
- 325×300×150 mm
- 325×300×150 mm
- W×D×H:
- 355×325×305 mm
- 355×325×305 mm
Frequently Bought Together