Order Entry
Austria
ContactUsLinkComponent
VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
Catalog # 85669.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
Catalog # 85669.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Colour:
    Neutral
  • Description:
    Ph Eur N 4000600
  • pH value:
    3,5

Specifications

About this item

Contents: ammonium acetate R / water R / hydrochloric acid R1.

  • Ph Eur N 4000600

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.

Ordering information: Normally manufactured to order. Please check with customer services.
Delivery: Delivery with certificate of analysis.