Order Entry
Austria
ContactUsLinkComponent
VWR®, Standard solution (250 ppm NH₄) for the preparation of Ammonium standard solution (2.5 ppm NH₄) Reag.Ph.Eur.; 5000301
VWR®, Standard solution (250 ppm NH₄) for the preparation of Ammonium standard solution (2.5 ppm NH₄) Reag.Ph.Eur.; 5000301
Catalog # 88084.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Standard solution (250 ppm NH₄) for the preparation of Ammonium standard solution (2.5 ppm NH₄) Reag.Ph.Eur.; 5000301
Catalog # 88084.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    250 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Ammonium standard solution

Specifications

About this item

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.