Order Entry
Austria
ContactUsLinkComponent
Standard solution (200 ppm Fe) for the preparation of iron standard solution (20 ppm Fe) Reag. Ph. Eur.; 5001600
Standard solution (200 ppm Fe) for the preparation of iron standard solution (20 ppm Fe) Reag. Ph. Eur.; 5001600
Catalog # 88086.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Standard solution (200 ppm Fe) for the preparation of iron standard solution (20 ppm Fe) Reag. Ph. Eur.; 5001600
Catalog # 88086.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    200 mg/L
  • Matrix:
    2% H₂SO₄
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.