Order Entry
Austria
ContactUsLinkComponent
VWR®, Standard solution (1000 ppm SO₄) for the preparation of sulphate standard solution (100 ppm SO₄) Reag.Ph.Eur. 5002802
VWR®, Standard solution (1000 ppm SO₄) for the preparation of sulphate standard solution (100 ppm SO₄) Reag.Ph.Eur. 5002802
Catalog # 88090.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Standard solution (1000 ppm SO₄) for the preparation of sulphate standard solution (100 ppm SO₄) Reag.Ph.Eur. 5002802
Catalog # 88090.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Standard solution (1000 ppm SO₄)
  • Contents:
    K₂SO₄ in H₂O

Specifications

About this item

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.